Learn More
Zebra Technologies Corporation Label, Polyester, 4x6in (101.6x152.4mm); TT, Z-Ultimate 2000T White, Value Coated, Permanent Adhesive, 3in (76.2mm) core, 960/roll, 4/box, Plain
Supplier:  Zebra Technologies Corporation 10008518
Encompass_Preferred
This item has a minimum order quantity of 4 per supplier requirements
Label, Polyester, 4x6in (101.6x152.4mm); TT, Z-Ultimate 2000T White, Value Coated, Permanent Adhesive, 3in (76.2mm) core, 960/roll, 4/box, Plain
Catalog No. NC3256810