Learn More
GA International Transparent Cryogenic Labels for Thermal-Transfer Printers 2" x 2" / 50.8mm x 50.8mm Rectangle Transparent From -196°C to +100°C (-321°F to +212°F)
Supplier:  GA International HBCL27NOTC31
Encompass_Preferred
Transparent thermoplastic cryogenic labels for bio-medical research and clinical laboratories. Withstand long-term storage in deep-freeze and harsh laboratory environments, including liquid and vapor phase liquid nitrogen* (-196°C/-321°F), lab freezers (-80°C/-112°F), and dry ice transportation. Ideal for labeling vials where an unobstructed view of contents is required. Their high temperature and water resistance make these labels perfectly suitable for heated water bath thawing applications. When printed in mirror-image mode, these labels can be applied to the underside of clear plates for easy reading.
Catalog No. 50-005-70965