Learn More
GA International Permanent Solvent-Resistant Color Labels for Containers 3" x 2" / 76.2mm x 50.8mm Rectangle Yellow From -80°C to +100°C (-112°F to +212°F)
Supplier:  GA International AUC56C10.5YE
Encompass_Preferred
Solvent and chemical-resistant permanent color labels for thermal-transfer barcode printers. Ideal for color coding different containers exposed to harsh chemicals and solvents such as xylene, toluene, methyl ethyl ketone (MEK), acetone, thinners, commercial cleaners, and window cleaners. These extra-durable labels can be printed with our XAR-class solvent-resistant ribbons to obtain a chemical-resistant printout.
Catalog No. 50-005-71416