Learn More
Research Products International Corp LABeler, Standard Label Tape Cassette, 12mm, 26 Feet, White with Black Print
SDP
Supplier:  Research Products International Corp 165103
Encompass
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -50°C.
Catalog No. 50-136-7668