Learn More
Essendant LABELMAKERRHINO5200YL
Supplier:  Essendant DYM1755749
Encompass_Preferred
Designed for professional installers in real-world jobsite conditions Quickly creates crystal-clear professionally formatted labels Exclusive hot keys automatically size space and format labels for wires and cables terminal blocks 110 blocks electrical and patch panels vertical and fixed-length applications Pre-programmed symbol library puts industry symbols punctuation currency and fractions at your fingertips Prints Code 39 and Code 128 bar codes on 3/4 labels Large two-line display is backlit Slip-resistant grip and rubber bumper protect against damage Lanyard loop provides the option of hands-free portability Uses industrial strength labels that resist extreme heat UV rays moisture chemicals and solvents Automatic shut-off maximizes battery life Uses six AA batteries or quick-charging lithium battery (all sold separately) Power Source(s)- Li-ion Battery (Sold Separately) Tape Size- 0 25 0 38 0 5 0 75 Number of Printed Lines- 5 Display CharactersxDisplay
Catalog No. 50-000-22363