Learn More
Andwin Scientific Laser Tough-Tags 0.94 x 0.50" 2,975/pk, Blue
SDP
Supplier:  Andwin Scientific TTSW2240B
Encompass_Preferred
Laboratory labels formatted for laser printers. Tough-Tags are chemically inert laboratory labels that strongly adhere to microtubes, test tubes, storage boxes and microscope slides. They resist organic solvents and caustic agents. Tough-Tags are perfect for accurately identifying important samples stored in moderate freezer temperatures.
Catalog No. NC1660858