Learn More
TARGETMOL CHEMICALS INC Labetalol 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T20570100MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Labetalol (Apo-Labetalol) is a mixed alpha/beta-adrenergic antagonist.It is used to treat high blood pressure. It can be given by mouth for long term hypertension management or intravenously in severe hypertensive situations. The pharmacokinetics of labetalol is affected by the common CYP2C19 polymorphisms. Purity 99.89%
Catalog No. 50-287-7774