Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Labetalol | 36894-69-6 | 99.9% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY12138310MG

Encompass_Preferred

Labetalol is an antihypertensive α1- and non-selective β-adrenergic receptor antagonist supplied as a solid analytical standard for research and reference use in pharmacology and medicinal chemistry.

  • Used as an analytical standard and research reagent.
  • CAS 36894-69-6; molecular formula C19H24N2O3; molecular weight 328.41.
  • High purity (99.94% by supplier specification).
  • Solid form, white to off-white powder; pack size 10 MG.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-30677


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.