Learn More
Medchemexpress LLC Labetalol hydrochloride | 32780-64-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1108R5MG
Encompass_Preferred
Labetalol (AH5158) hydrochloride is an orally available, selective α1-adrenergic receptor and non-selective β-adrenergic receptor competitive antagonist. It is an antihypertensive molecule that partially crosses the blood-brain barrier and has little effect on cardiac output. Labetalol hydrochloride can be used in the study of cardiovascular diseases, such as hypertension during pregnancy.
  • Orally available selective α1-adrenergic receptor and non-selective β-adrenergic receptor competitive antagonist.
  • Antihypertensive molecule.
  • Partially crosses the blood-brain barrier.
  • Has little effect on cardiac output.
  • Can be used in the study of cardiovascular diseases, including hypertension during pregnancy.
Catalog No. 50-004-13069