Learn More
Medchemexpress LLC Labetalol (hydrochloride) | 32780-64-6 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY128379
Encompass_Preferred
Labetalol hydrochloride is an orally available, selective α1-adrenergic receptor and non-selective β-adrenergic receptor competitive antagonist. It is an antihypertensive molecule that partially crosses the blood-brain barrier and has little effect on cardiac output. It can be used in the study of cardiovascular diseases, such as hypertension during pregnancy.
  • Orally available
  • Selective α1-adrenergic receptor and non-selective β-adrenergic receptor competitive antagonist
  • Antihypertensive molecule
  • Partially crosses the blood-brain barrier
  • Has little effect on cardiac output
  • Used in the study of cardiovascular diseases, such as hypertension during pregnancy
  • Exhibits greater affinity for β-adrenergic sites
  • Blocks β-adrenergic responses at low doses
  • Blocks α-adrenergic responses at high doses
Catalog No. 50-003-42983