Learn More
TARGETMOL CHEMICALS INC Labetalol hydrochloride 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0348500MG
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Labetalol hydrochloride (Sch-15719W), a salicylamide derivative, is a non-cardioselective blocker of beta-adrenergic receptors and alpha1-adrenergic receptors. Purity 99.85%
Catalog No. 50-288-0762