Learn More
Cayman Chemical LabetalolhydrochlorIde 5g
SDP
Supplier:  Cayman Chemical 202495 G
Encompass_Preferred
A competitive antagonist of both α- and β-adrenoreceptors that is effective at nanomolar concentrations
Catalog No. 50-298-4934