Learn More
Medchemexpress LLC Labetuzumab, 1mg, | CAS No. : 219649-07-7
SDP
Supplier:  Medchemexpress LLC HYP991601MG
Encompass_Preferred
Labetuzumab is a humanised anti-carcinoembryonic antigen (CEA) monoclonal antibody that inhibits tumour growth and sensitises human medullary thyroid cancer xenografts to Dacarbazine chemotherapy.
Catalog No. NC3361504