Learn More
Medchemexpress LLC Labetuzumab govitecan, 1mg, (Synonyms: IMMU 130; hMN-14-SN-38) | CAS No. : 1469876-18-3
SDP
Supplier:  Medchemexpress LLC HYP996811MG
Encompass_Preferred
Labetuzumab govitecan (IMMU 130) is an Anti-CEACAM5/SN-38 antibody-drug conjugate (ADC). Labetuzumab govitecan consists of the antibody Labetuzumab, SN-38, and a pH-sensitive Linker. Labetuzumab govitecan can be used for research of cancers, such as colorectal cancer.
Catalog No. NC3361528