Learn More
Hach Company Hach TU5200 Laboratory Laser Turbidimeter, EPA Version (LPV442.99.01012)
Supplier: Hach Company LPV442.99.01012
The Hach TU5200 Laboratory Laser Turbidimeter (TU5 Series) is a benchtop instrument engineered for precise, EPA-compliant turbidity measurements. It employs patented 360° x 90° Detection technology to maximize scattered light collection, improving low-level sensitivity and repeatability while eliminating the need for silicone oil and vial indexing.
- Provides EPA-compliant measurement range of 0-700 NTU and conversions to FNU, FTU, mg/L, and EBC.
- Delivers high accuracy: ±2% plus 0.01 NTU (0-40 NTU) and ±10% of reading (40-1000 NTU).
- Uses a 650 nm laser wavelength for stable, low-level detection.
- Accepts standard sample vials; no oil or indexing required.
- Compact benchtop design with integrated color display and simple operation.
- Includes stabilizing standards kit, sample vials, power supply, dust cover, and vial stand.
- Operates on 100-240 VAC, 50/60 Hz for global compatibility.