Learn More
Electron Microscopy Sciences Lab-Tek II CC2 Slide, 2 Chamber, 12 Trays
SDP
Supplier:  Electron Microscopy Sciences 7036026
Encompass_Preferred
Lab-Tek II Chamber Slide products offer optimal surface for excellent attachment and growth of cells, easily distinguished by the blue mark. This design allows for the removal of the media chamber and adhesive from the slide, using a separation device. The blue hydrophobic border is raised slightly and isolates the individual wells, and prevents cross contamination. The slide can be coversliped.
Chamber Slide products are sterile and have removable, polystyrene media chambers. Glass microscope slides are low fluorescent (25 x 75 x 1.2 mm) with rounded corners, SuperFrost printed area, and inert hydrophobic well border printed on slide. The adhesive is biocompatible. Polystyrene covers. Glass surface is chemically modified for growing fastidious cells.
Catalog No. 50-189-9251