Learn More
Andwin Scientific LABTONE GP CLEANER 8/LBS
SDP
Supplier:  Andwin Scientific 89030112
Encompass_Preferred
The Labtone Laboratory Cleaning Compound is ideal for cleaning laboratory glasswares. This compound contains carbonates, phosphates, and organic detergent in balanced proportions. This product eliminates dirt, grease, media, and soil with no scrubbi
Catalog No. NC9599600