Learn More
Medchemexpress LLC Lacidipine | 103890-78-4 | 99.40% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0347
Encompass_Preferred
Lacidipine is an orally active and highly selective L-type calcium channel blocker that acts on smooth muscle calcium channels, primarily dilates peripheral arteries, reduces peripheral resistance, and has long-lasting anti-hypertensive activity. It protects HKCs from apoptosis induced by ATP depletion and recovery by modulating the caspase-3 pathway. This product is for research use only and can be used in studies of hypertension, atherosclerosis, and acute kidney injury (AKI).
  • Orally active and highly selective L-type calcium channel blocker
  • Primarily dilates peripheral arteries and reduces peripheral resistance
  • Has long-lasting anti-hypertensive activity
  • Protects HKCs from apoptosis by modulating the caspase-3 pathway
  • Useful in studies of hypertension, atherosclerosis, and acute kidney injury (AKI)
Catalog No. 50-004-07063