Learn More
Medchemexpress LLC Lacritin Human His 10ug

Supplier: Medchemexpress LLC HYP7091110UG
Recombinant human Lacritin (A19-A138) with an N-terminal 6xHis tag, supplied as a lyophilized powder for research use. The protein is expressed in E. coli, purified to high purity with low endotoxin, and provided with storage and reconstitution guidance for biochemical and cell-based studies.
- Recombinant fragment A19-A138 with N-terminal 6xHis tag.
- Expressed in E. coli for efficient production.
- Purified to greater than 95% for reliable analytical and functional assays.
- Lyophilized formulation for extended shelf life and convenient storage.
- Low endotoxin level (<1 EU/μg) suitable for many cell-based applications.
- Reconstitution recommended at ≥100 μg/mL; aliquot and store at -20 °C or -80 °C.
- Approximately 23.0 kDa molecular weight for analytical confirmation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.