Learn More
Enzo Life Sciences Lactacystin, (200µg), CAS Number: 133343-34-7
SDP
Supplier:  Enzo Life Sciences BMLPI1040200
Encompass_Preferred
Key inhibitor for use in proteasome research. Formula: C15H24N2O7S. MW: 376.4. CAS Number: 133343-34-7. Purity: ≥97% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water (10mg/ml), 100% ethanol (1mg/ml), methanol, DMSO (20mg/ml), dimethyl formamide (20mg/ml) or acetonitrile (20mg/ml). Use/Stability: Organic stock solutions are stable for 3 months when stored at -20°C. Subject to hydrolysis in aqueous buffers. We do not recommend storing aqueous solutons for more than one day. Long Term Storage: -20°C
Catalog No. 50-103-1784