Learn More
Medchemexpress LLC Lactic acid-d4 sodium | 344299-52-1 | 99.0% | 116.08 g/mol | C3HD4NaO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB2227BS10MG
Encompass_Preferred
Lactate-d4 sodium is the deuterium-labeled sodium salt of lactic acid provided as a 60% w/w aqueous solution for use as an isotopic tracer and analytical standard. The reagent is intended for research applications in metabolomics and biochemical studies and is characterized by high isotopic/purity and a molecular weight of 116.08 g/mol (CAS 344299-52-1).
  • Deuterium-labeled endogenous metabolite for tracer and quantitation studies.
  • Supplied as a 60% w/w aqueous solution for ease of handling.
  • High isotopic/purity suitable for mass spectrometry and analytical workflows.
  • Applicable to studies of lactate metabolism, HCAR1 signaling, and lysine lactylation.
  • Available in small pre-weighed quantities for research use.
Catalog No. 50-002-97320