Learn More
ATCC Lactiplantibacillus plantarum
Supplier:  ATCC 10241
Encompass
Lactiplantibacillus plantarum strain [BUCSAV 248, NCDO 343, NCIB 7220, NCIB 8184, NCTC 7220] is a whole-genome sequenced bacterium that was isolated in England from Sauerkraut. This strain produces acetylcholine.
Catalog No. 50-238-2412
 Restricted Item