Learn More
Medchemexpress LLC Lacto-N-biose I (Galβ1-3GlcNAc) | 50787-09-2 | 98.0% | 383.35 g/mol | C14H25NO11 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1414885MG
Encompass_Preferred
Lacto-N-biose I (Galβ1-3GlcNAc) is a naturally occurring disaccharide glycan and an endogenous metabolite used in glycobiology research. It functions as an acceptor substrate for fucosyltransferase enzymes (for example, α1,2-fucosyltransferase from Helicobacter pylori) and is applied in enzymatic assays, glycan synthesis studies, and investigations of enzyme substrate specificity.
  • Used as an acceptor substrate for fucosyltransferase assays.
  • High purity (98.0%), suitable for biochemical applications.
  • Available in small research quantities for assay development.
  • Supports studies of glycan biosynthesis and enzyme specificity.
  • Compatible with enzymatic and analytical workflows.
Catalog No. 50-002-22784