Learn More
Cayman Chemical Lacto-N-tetraose 10mg
SDP
Supplier:  Cayman Chemical 3903410 MG
Encompass_Preferred
An oligosaccharide; promotes the growth of the bacterium B. longum and B. breve; levels are increased in breast milk from mothers of rotavirus-positive neonates with gastrointestinal symptoms and are positively correlated with levels of Enterobacter and Klebsiella in maternal milk and infant stool
Catalog No. 50-291-8254