Learn More
ATCC Lactobacillus acidophilus
Supplier:  ATCC 53544
Encompass
Lactobacillus acidophilus strain BT1005 is a bacterium that was isolated from the rectal swab of a human infant. This strain is cited to have applications in the production of animal feed dietary supplements.
Catalog No. 50-238-1258