Learn More
ATCC Lactobacillus fermentum; Strain: L888
Supplier:  ATCC 14932
Encompass
Lactobacillus fermentum strain L888 was isolated from saliva. This bacterial strain is grown aerobically in lactobacilli MRS medium.
Catalog No. 50-238-3425