Learn More
Alkali Scientific Lactobacillus MRS Broth Powder, 500 G
SDP
Supplier:  Alkali Scientific OBA003CP
Encompass_Preferred
Lactobacillus MRS Broth Powder is a specialized medium designed for the selective growth of Lactobacillus species, available in a 500g package. This nutrient-rich broth contains a combination of amino acids, vitamins, and minerals that promote the growth and fermentation of Lactobacillus. It is used in microbiology laboratories for studying probiotics, fermentation processes, and dairy production. The medium is formulated to support the rapid growth of Lactobacillus and similar bacteria, making it essential in industrial and research settings focused on probiotic development, dairy products, and microbial fermentation. Its versatile formulation also supports a variety of applications in food and beverage production.
Catalog No. 50-006-61297