Learn More
Medchemexpress LLC Lactobionic acid | 96-82-2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYN705910G
Encompass_Preferred
Lactobionic acid is a bionic acid naturally found in Caspian Sea yogurt, composed of a gluconic acid bonded to a galactose. It offers antioxidant, antimicrobial, chelating, stabilizer, acidulant, and moisturizing properties. Utilized in foodstuffs for new functional products and to combat food-borne pathogens, it also inhibits DNA repair and protein synthesis, induces oxidative stress, and inhibits metabolic pathways against MRSA.
  • Possesses antioxidant, antimicrobial, chelating, stabilizer, acidulant, and moisturizing properties
  • Utilized in foodstuffs for new functional products and to combat food-borne pathogens
  • Inhibits DNA repair and protein synthesis
  • Induces oxidative stress
  • Inhibits metabolic pathways against MRSA
  • Negatively affects growth of *Staphylococcus aureus*, damaging cell membrane integrity
  • Leads to cell damage in *P. taetrolens* cells
Catalog No. 50-004-19672