Learn More
Medchemexpress LLC Lactonic sophorolipid | 148409-20-5 | 97.3% | 688.80 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY137371
Encompass_Preferred
Lactonic sophorolipid acts as an apoptosis inducer and antimicrobial surfactant with antitumor activity. It regulates Bax/Bcl-gene expression via caspase-3/9, promoting apoptosis in tumor cells. It also disrupts cell membrane permeability for antibacterial effects (MIC 100-400 μg/mL for oral pathogens) and promotes mitochondrial membrane potential depolarization, activating the intrinsic apoptotic pathway. It can synergize with antibiotics and is used in liver cancer research and oral hygiene antibacterial agent development.
  • Induces apoptosis in tumor cells
  • Regulates Bax/Bcl-gene expression
  • Disrupts cell membrane permeability
  • Exerts antibacterial effects
  • Promotes mitochondrial membrane potential depolarization
  • Activates the intrinsic apoptotic pathway
  • Synergizes with antibiotics
  • Inhibits HepG2 cell viability
  • Inhibits proliferation in colon cancer cells
  • Disrupts biofilm formation
Catalog No. 50-003-51007