Learn More
Medchemexpress LLC Lactose | 63-42-3 | 98.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB2123500MG
Encompass_Preferred
Lactose is a β-galactoside composed of galactose and glucose residues, and it is the primary carbohydrate found in mammalian breast milk. As a macronutrient and an inducer of host innate immune responses, Lactose exhibits immune modulatory functions.
  • Inhibits regulatory T-cell-mediated suppression of effector T-cell interferon-γ and IL-17 production.
  • Improves acute pancreatitis in studies.
  • Alleviated the severity of caerulein-induced acute pancreatitis (AP) in mice.
  • Modulated early immune cell infiltration into the pancreas and pancreatic cytokine/chemokine production.
  • Suppressed ERK1/2 and NF-κB p65 signaling to inhibit cytokine production in pancreatic acinar cells.
Catalog No. 50-004-09682