Learn More
YSI 3P LifeScience 2705 Lactose Buffer (8 vials - 4 liters)
Supplier:  YSI 3P LifeScience 2705
Encompass
Buffer for Glycerol, Lactose or Galactose. 8 pouches. Reconstitute to make a total of 4 liters.
Catalog No. NC9334033