Learn More
Medchemexpress LLC α-Lactose (hydrate) | 5989-81-1 | MFCD00150747 | 98.0% | 360.31 g/mol | C12H22O11·H2O | 500 G
SDP
Supplier:  Medchemexpress LLC HYW087904500G
Encompass_Preferred
α-Lactose (hydrate) is the alpha anomer of lactose supplied as the monohydrate crystalline form. It is used as a pharmaceutical excipient, diluent, bulking agent for tablet formulations, and as a carrier or filler in research reagents.
  • Monohydrate crystalline form for consistent handling and dosing
  • Common pharmaceutical excipient and diluent
  • Suitable for tablet formulations and as a bulking agent
  • Used in food and biochemical research applications
  • CAS number 5989-81-1; molecular weight 360.31 g/mol
Catalog No. 50-002-08313