Learn More
Medchemexpress LLC Lactose (Standard) | 63-42-3 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB2123R100MG
Encompass_Preferred
Lactose (Standard) is an analytical standard of Lactose, intended for research and analytical applications. It is a β-galactoside consisting of galactose and glucose residues, commonly found as the main carbohydrate in mammalian breast milk. As a macronutrient and inducer of host innate immune responses, it possesses immune modulatory functions.
  • Used as an analytical standard.
  • Supports qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS applications.
Catalog No. 50-004-11025