Learn More
Selleck Chemical LLC Lactulose S1511-1g
SDP
Supplier:  Selleck Chemical LLC S15111G
Encompass_Preferred
Lactulose a disaccharide composed of galactose and fructose is a stimulator of health-promoting bacterium growth in the gastrointestinal tract used to treat chronic constipation and hepatic encephalopathy
Catalog No. 50-311-1358