Learn More
Apexbio Technology LLC Lactulose 4618-18-2 50mg
SDP
Supplier:  Apexbio Technology LLC B177650
Encompass_Preferred
Lactulose (4618-18-2) is a synthetic non-absorbable disaccharide composed of galactose and fructose units It is designed to reach the colon intact due to the absence of digestive enzymes in the upper gastrointestinal tract where it undergoes bacterial fermentation to form short-chain fatty acids primarily lactic and acetic acid thereby lowering intraluminal pH and increasing osmotic water retention Lactulose exerts its biological activity primarily through colonic acidification and modulation of water content by promoting fermentation-derived metabolite production No IC50 values have been reported for lactulose Based on these pharmacological properties lactulose holds research potential in studies investigating constipation mechanisms ammonia detoxification and the pathophysiology of hepatic encephalopathy
Catalog No. 50-000-92609