Learn More
Medchemexpress LLC Laduviglusib trihydrochloride | 1782235-14-6 | MFCD29079432 | 99.1% | 574.72 | C22H21Cl5N8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10182B5MG
Encompass_Preferred
Laduviglusib trihydrochloride is the trihydrochloride salt of a potent, selective glycogen synthase kinase-3 (GSK-3) inhibitor supplied as a high-purity solid for research use. It is commonly used in cell signaling and preclinical studies to probe GSK-3-related pathways, including autophagy and Wnt/β-catenin signaling.
  • Potent, selective GSK-3 inhibitor with low-nanomolar activity.
  • Trihydrochloride salt form as a stable solid for handling.
  • High purity (99.1%) suitable for biochemical and cellular assays.
  • Useful for studies of autophagy and Wnt/β-catenin signaling pathways.
  • Supplied in small milligram quantities for research applications.
Catalog No. 50-002-48771