Learn More
Medchemexpress LLC Laduviglusib trihydrochloride | 1782235-14-6 | MFCD29079432 | 99.1% | 574.72 g/mol | C22H21Cl5N8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10182B100MG
Encompass_Preferred
Laduviglusib trihydrochloride is the trihydrochloride salt of laduviglusib (CHIR-99021), a potent and selective inhibitor of glycogen synthase kinase-3 (GSK-3α/β) used in biochemical and cellular research. The material is supplied as a dry powder with characterized purity and molecular weight for laboratory applications; it is for research use only and not for human or clinical use.
  • Potent, selective GSK-3α/β inhibition for pathway modulation.
  • High purity suitable for biochemical assays and cell studies.
  • Trihydrochloride salt form enhances solubility in polar solvents.
  • Confirmed molecular weight and structural identity.
  • Commonly used to promote stem cell self-renewal and activate Wnt/β-catenin signaling.
Catalog No. 50-002-45220