Learn More
Medchemexpress LLC Lagosin 5mg | 6834-98-6 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1066815MG
Encompass_Preferred
Lagosin (Fungichromin) is a polyene macrolide antibiotic Lagosin has demonstrated broad-spectrum antifungal activity and is impervious to agent resistance[1
Catalog No. 50-002-40961