Learn More
Cayman Chemical LaHydroxyglutarIc AcIdod 10mg
SDP
Supplier:  Cayman Chemical 1187610 MG
Encompass_Preferred
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; competitively inhibits α-ketoglutarate-dependent dioxygenases, including several involved in histone lysine and DNA demethylation
Catalog No. 50-295-2173