Learn More
Charles River Laboratories Kinetic Turbidimetric LAL Test Reagents, 0.03 EU/mL, Case of 100
Supplier:  Charles River Laboratories R15003BX
Encompass
Kinetic Turbidimetric LAL - Our kinetic turbidimetric LAL reagent, KTA, yields quantitative endotoxin values when used with a microplate equipped with endotoxin-measuring software. The reagent is buffered to provide significant interference-resistance properties and FDA-licensed for use in product release testing. With KTA, you will receive accurate endotoxin results on even your most difficult products in the same amount of time it takes to run a gel-clot assay. Our traditional reagent, KTA is suitable for both kinetic and gel-clot analyses and permits a direct correlation between methods.
Catalog No. NC9888726
Only null left
Add to Cart
Request a Quote