Learn More
Bioworld Lambda Broth, 100 g
Supplier:  Bioworld 306210051
Encompass_Preferred
Lambda BrothMedia for the propagation of lambda phageBiotechnology Grade pH: 6.9Dehydrated Appearance: light beige powder Soluble in water
Catalog No. 50-254-427