Learn More
Agilent Technologies LAMBDA CE6 INDUCTION KIT
Supplier:  Agilent Technologies 235200
Encompass
lambda CE6 Induction Kit, BL21 derivatives for tight expression control, express toxic proteins, use BL21, BL21-Gold or BL21-CodonPlus cells.
Catalog No. 50-125-361