Learn More
New England Biolabs, Inc. Lambda Exonuclease – 1000 units
SDP
Supplier:  New England Biolabs, Inc. M0262S
Encompass
Lambda Exonuclease is a processive enzyme that catalyzes 5' to 3' mononucleotide removal from dsDNA with a preferred substrate of a 5' phosphate. Ideal for conversion of linear double-stranded DNA to single-stranded DNA via preferred activity on 5'-phosphorylated ends.
Catalog No. 50-811-687