Learn More
ABclonal Technology LAMC1 Rabbit pAb
SDP
Catalog No. 5000105748
Encompass_Preferred
Laminins are extracellular matrix glycoproteins that form the major noncollagenous component of basement membranes, involved in processes like cell adhesion, differentiation, migration, and metastasis. They consist of three non-identical chains: alpha, beta, and gamma, forming a cruciform structure. Each chain is encoded by a distinct gene with multiple isoforms. Different combinations of alpha, beta, and gamma chains form various laminin isoforms, such as laminin 1 (alpha1beta1gamma1). The functions of these chains are not fully understood, though their tissue distribution suggests diverse roles. This gene encodes the gamma 1 chain isoform. Unlike the beta chain, it lacks a short alpha region. Inactivation of both gamma 1 chain alleles in transgenic mice causes a lack of basement membranes, showing its role in laminin assembly. Multiple polyadenylation sites generate two mRNA variants (5.5 and 7.5 kb) in humans.
Catalog No. 50-001-05748 Supplier ABclonal Technology Supplier No. A160201000UL
Laminins are extracellular matrix glycoproteins that form the major noncollagenous component of basement membranes, involved in processes like cell adhesion, differentiation, migration, and metastasis. They consist of three non-identical chains: alpha, beta, and gamma, forming a cruciform structure. Each chain is encoded by a distinct gene with multiple isoforms. Different combinations of alpha, beta, and gamma chains form various laminin isoforms, such as laminin 1 (alpha1beta1gamma1). The functions of these chains are not fully understood, though their tissue distribution suggests diverse roles. This gene encodes the gamma 1 chain isoform. Unlike the beta chain, it lacks a short alpha region. Inactivation of both gamma 1 chain alleles in transgenic mice causes a lack of basement membranes, showing its role in laminin assembly. Multiple polyadenylation sites generate two mRNA variants (5.5 and 7.5 kb) in humans.