Learn More
Medchemexpress LLC Lamivudine | 134678-17-4 | 99.9% | 500 MG
Supplier: Medchemexpress LLC HYB0250500MG
Lamivudine (BCH-189) is an orally active nucleoside reverse transcriptase inhibitor (NRTI). It is capable of inhibiting both HIV reverse transcriptase 1/2 and the reverse transcriptase of hepatitis B virus. Additionally, lamivudine salicylate has the ability to penetrate the central nervous system (CNS).
- Orally active nucleoside reverse transcriptase inhibitor (NRTI)
- Inhibits HIV reverse transcriptase 1/2
- Inhibits reverse transcriptase of hepatitis B virus
- Salicylate form can penetrate the CNS