Learn More
Medchemexpress LLC Lamotrigine-13C3 | 1188265-38-4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0495S41MG
Encompass_Preferred
Lamotrigine-13C3 is the 13C-labeled version of Lamotrigine, a potent and orally active anticonvulsant or antiepileptic agent. It selectively blocks voltage-gated Na+ channels, stabilizes presynaptic neuronal membranes, and inhibits glutamate release. This product is suitable for research on epilepsy, focal seizure, and similar conditions.
  • 13C-labeled Lamotrigine, suitable as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent and orally active anticonvulsant/antiepileptic agent
  • Selectively blocks voltage-gated Na+ channels
  • Stabilizes presynaptic neuronal membranes
  • Inhibits glutamate release
Catalog No. 50-004-09285