Learn More
Medchemexpress LLC Lamotrigine-13C3,d3 | 1246815-13-3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0495S1MG
Encompass_Preferred
Lamotrigine-13C3,d3 is a 13C-labeled version of Lamotrigine, a potent and orally active anticonvulsant or antiepileptic agent. It functions by selectively blocking voltage-gated Na+ channels, stabilizing presynaptic neuronal membranes, and inhibiting glutamate release.
  • Useful for research related to epilepsy and focal seizures
  • Incorporates stable heavy isotopes as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08874