Learn More
Medchemexpress LLC Lamotrigine | 84057-84-1 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB04955G
Encompass_Preferred
Lamotrigine is a potent, orally active anticonvulsant and antiepileptic agent. It selectively blocks voltage-gated Na+ channels, stabilizing presynaptic neuronal membranes and inhibiting glutamate release. This compound is suitable for research into epilepsy, focal seizures, and similar conditions.
  • Potent, orally active anticonvulsant
  • Selectively blocks voltage-gated Na+ channels
  • Stabilizes presynaptic neuronal membranes
  • Inhibits glutamate release
  • Used in research for epilepsy and focal seizure
Catalog No. 50-004-05270