Learn More
Optimize Technologies Deuterium Lamp, Perkin-Elmer 200 UV/VIS
SDP
Supplier:  Optimize Technologies 333203117
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number N2920149.
Catalog No. 50-200-6645