Learn More
MSC BluShield Adjustable Spray Nozzle and Wand, Brass, 21 in, 3000 psi
Supplier:  MSC 63959977
Encompass
This adjustable spray nozzle and wand is designed for pressure washers with capacities up to 3000 PSI. It provides adjustable spray patterns from precise pin-point to expansive wide fan (0-40 degrees) and seamlessly transitions from high to low pressure for applying cleaning solutions. This brass assembly connects to spray gun fittings, offering universal compatibility.
  • Designed for pressure washers up to 3000 PSI
  • Adjustable spray patterns from 0 to 40 degrees (pin-point to wide fan)
  • Seamlessly transitions from high to low pressure for applying cleaning solutions
  • Connects effortlessly to spray gun fittings for universal compatibility
Catalog No. 50-283-8794